Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKD3/nPKC nu Antibody (OTI3D8), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273595A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273595A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273595A405 Supplier Novus Biologicals Supplier No. NBP273595AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRKD3/nPKC nu Monoclonal specifically detects PRKD3/nPKC nu in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PRKD3/nPKC nu
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3D8
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.13, EPK2PKD3, nPKC-nu, PKC-NU, PRKCNnPKC-NU, Protein kinase C nu type, protein kinase C, nu, protein kinase D3, Protein kinase EPK2, protein-serine/threonine kinase, serine/threonine-protein kinase D3
Gene Symbols PRKD3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 300-508 of human PRKD3/nPKC nu (NP_005804) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Diabetes Research, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23683.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.