Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRL-2/PTP4A2 Antibody (015), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB070627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070627 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070627 Supplier Novus Biologicals Supplier No. NBP290172IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PRL-2/PTP4A2 Monoclonal specifically detects PRL-2/PTP4A2 in Human samples. It is validated for ELISA.

Specifications

Antigen PRL-2/PTP4A2
Applications ELISA
Classification Monoclonal
Clone 15
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.48, HU-PP-1HH13, OV-1PTP4A, PRL-2HH7-2, PRL2protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA 2, protein tyrosine phosphatase type IVA, member 2, Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, PTP(CAAXII), PTPCAAX2phosphatase of regenerating liver 2, ptp-IV1a, ptp-IV1b
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PRL-2/PTP4A2 (Q12974-1; Asn2-Gln167).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8073
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.