Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRLH Polyclonal Antibody, Invitrogen™

Catalog No. PIPA571788
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571788 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571788 Supplier Thermo Scientific Supplier No. PA571788

Rabbit Polyclonal Antibody

Stimulates prolactin (PRL) release and regulates the expression of prolactin through its receptor GPR10. May stimulate lactotrophs directly to secrete PRL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRLH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Prlh
Gene Alias Prolactin-releasing peptide; PrRP; Prolactin-releasing hormone; Prolactin-releasing peptide PrRP31; Prolactin-releasing peptide PrRP20; PRLH; PRH
Gene Symbols Prlh
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 27-56 amino acids from the Central region of human PRLH
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 623503
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.