Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT1 Antibody (PRMT1-171) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12012189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12012189 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12012189 Supplier Novus Biologicals Supplier No. NB12012189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRMT1 Monoclonal specifically detects PRMT1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen PRMT1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone PRMT1-171
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Gene Alias ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1
Gene Symbols PRMT1
Host Species Mouse
Immunogen Monoclonal Anti-PRMT1 is derived from the PRMT1-171 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with full length PRMT1.
Purification Method Protein A or G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3276
Test Specificity This reacts with PRMT1 and does not cross-react with PRMT2, 3, 4, and 5.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.