Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT3 Antibody (RAB-C372), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB340458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340458 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340458 Supplier Novus Biologicals Supplier No. NBP320143MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)

Specifications

Antigen PRMT3
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Mass Spectrometry
Classification Monoclonal
Clone RAB-C372
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10196
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.