Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT5 Antibody (2H4), Novus Biologicals™
SDP

Catalog No. NB298243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298243 100 μL
NB298242 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298243 Supplier Novus Biologicals Supplier No. NBP326367100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen PRMT5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2H4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PRMT5 [UniProt O14744]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
Primary or Secondary Primary
Gene ID (Entrez) 10419
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.