Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Antibody (2H4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | PRMT5 |
---|---|
Clone | 2H4 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
PRMT5 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
A synthesized peptide derived from Human PRMT5 [UniProt O14744] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2H4 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
10419 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title