Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219935 Supplier Novus Biologicals Supplier No. NBP219935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRMT5 Polyclonal antibody specifically detects PRMT5 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).

Specifications

Antigen PRMT5
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation PBS (pH 7), 20% Glycerol, 1% BSA with 0.025% Proclin 300
Gene Accession No. O14744
Gene Alias EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
Gene Symbols PRMT5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRMT5. The exact sequence is proprietary.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
Primary or Secondary Primary
Gene ID (Entrez) 10419
Test Specificity Human. Species reactivity based on sequence identity: Mouse (98%), Pig (98%), Rat (97%), Bovine (98%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.