Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT7 Antibody (PCRP-PRMT7-1A7), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB187575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187575 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187575 Supplier Novus Biologicals Supplier No. NBP314145UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRMT7 Monoclonal antibody specifically detects PRMT7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen PRMT7
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-PRMT7-1A7
Conjugate DyLight 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7, EC 2.1.1, EC 2.1.1.125, EC 2.1.1.126, FLJ10640, Histone-arginine N-methyltransferase PRMT7, KIAA1933EC 2.1.1.-, myelin basic protein-arginine N-methyltransferase, protein arginine methyltransferase 7, protein arginine N-methyltransferase 7
Host Species Mouse
Immunogen Recombinant full-length human PRMT7 protein (Uniprot: Q9NVM4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54496
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.