Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proapoptotic Caspase Adaptor Protein Antibody (022), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB333678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333678 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333678 Supplier Novus Biologicals Supplier No. NBP290320APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Proapoptotic Caspase Adaptor Protein Monoclonal antibody specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Proapoptotic Caspase Adaptor Protein
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 022
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Proapoptotic Caspase Adaptor Protein (Uniprot#: Q8WU39-1, Met 1- Thr 185)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 51237
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.