Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proapoptotic Caspase Adaptor Protein Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB100917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100917 Supplier Novus Biologicals Supplier No. NBP297099UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proapoptotic Caspase Adaptor Protein Polyclonal specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Proapoptotic Caspase Adaptor Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Proapoptotic Caspase Adaptor Protein (Q8WU39-1; Met 1- Thr 185).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 51237
Test Specificity Human MZB1/PERP1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.