Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procaine hydrochloride, 100.3%, MP Biomedicals™

Catalog No. icn19026050 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 g
100 g
250 g

Catalog No. ICN19026050

Supplier: MP Biomedicals, Inc 0219026050

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
PubChem CID 5795
ChEBI CHEBI:8431
IUPAC Name hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Assay Percent Range 1.003
Quantity 50 g
Formula Weight 272.8
Physical Form Crystalline
Chemical Name or Material Procaine hydrochloride
EINECSNumber 200-077-2
Recommended Storage Store at Room Temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.