Learn More
Description
Specifications
Specifications
| Antigen | Procalcitonin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 38F11 |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump |
| Gene Symbols | CALCA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
