Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRODH Antibody, Novus Biologicals™
SDP

Catalog No. NB1002832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002832 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002832 Supplier Novus Biologicals Supplier No. NB1002832
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PRODH Polyclonal specifically detects PRODH in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen PRODH
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:4000, Immunohistochemistry 6-7ug/ml, Immunohistochemistry-Paraffin 6-7ug/ml
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O43272
Gene Alias EC 1.5.99.8, FLJ33744, HSPOX2, mitochondrial, p53-induced gene 6 protein, PIG6MGC148078, POX2, PRODH2MGC148079, proline dehydrogenase (oxidase) 1, proline dehydrogenase (proline oxidase ), Proline oxidase, proline oxidase, mitochondrial, SCZD4, tumor protein p53 inducible protein 6
Gene Symbols PRODH
Host Species Goat
Immunogen Peptide with sequence C-ERDGSGTNKRDKQYQ corresponding to internal region according to NP_057419.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5625
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.