Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proenkephalin Antibody, Novus Biologicals™
SDP

Catalog No. NBP120963 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120963 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120963 Supplier Novus Biologicals Supplier No. NBP120963
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Proenkephalin Polyclonal specifically detects Proenkephalin in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Peptide ELISA.

Specifications

Antigen Proenkephalin
Applications Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 0.02 - 0.05 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001002927.1
Gene Alias enkephalin A, preproenkephalin, proenkephalin, proenkephalin-A
Gene Symbols PENK
Host Species Goat
Immunogen Peptide with sequence C-YKDSSKQDESH corresponding to internal region according to Uniprot Mouse NP_001002927.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5179
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.