Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Profilin 2 Antibody (010), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB038544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038544 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038544 Supplier Novus Biologicals Supplier No. NBP290284DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Profilin 2 Monoclonal specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Profilin 2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias D3S1319E, PFL, profilin 2, Profilin II, profilin-2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Profilin 2 (AAH18049.1; Met1-Phe140).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.