Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Profilin 2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB328991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328991 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328991 Supplier Novus Biologicals Supplier No. NBP300301MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Profilin 2 Polyclonal antibody specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Profilin 2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias D3S1319E, PFL, profilin 2, Profilin II, profilin-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Profilin 2 (Accession#: AAH18049.1; Met1-Phe140)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5217
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.