Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Profilin 3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
| Antigen | Profilin 3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5121476
|
Invitrogen™
PA5121476 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5121475
![]() |
Invitrogen™
PA5121475 |
100 μL |
Each for $511.50
|
|
|||||
Description
PFN3 (Profilin 3) is a Protein Coding gene. Among its related pathways are Association Between Physico-Chemical Features and Toxicity Associated Pathways. Gene Ontology (GO) annotations related to this gene include actin binding and lipid binding. An important paralog of this gene is PFN1. The product of this gene belongs to the profilin family of proteins. This protein binds to actin and affects the structure of the cytoskeleton. It also may be involved in spermatogenesis. It is a single exon gene.Specifications
| Profilin 3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P60673 | |
| 345456 | |
| Met1-Arg133 (Accession P60673), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PFN3 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Profilin III | |
| PFN3 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title