Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 500 Elbow Support

Catalog No. 19035285D Shop All Ergodyne Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-035-285D Medium
19-035-285E Large
19-035-285F X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-035-285D Supplier Ergodyne™ Supplier No. 16003
Add to Cart
Add to Cart

Ideal for jobs requiring repetitive arm motions

  • Wide base of support directs pressure away from tendons for improved performance with maximum comfort
  • Fully adjustable for a secure, exact fit
  • Wear over clothing or next to skin
  • Machine washable
  • Black

Specifications

Color Black
Product Type Elbow Support
Design Contoured
Size Medium
For Use With (Application) Jobs requiring repetitive arm motions
Hand Ambidextrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.