Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Antibody (11P14), Alexa Fluor™ 750
SDP

Catalog No. N102795A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102795A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102795A750 Supplier Novus Biologicals Supplier No. NB1002795AF750

Mouse Monoclonal Antibody

Progesterone Monoclonal specifically detects Progesterone in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Progesterone
Applications Western Blot, ELISA
Classification Monoclonal
Clone 11P14
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 4-Pregnen-3,20-dione
Host Species Mouse
Immunogen 11a-Hydroxyprogesterone conjugated to bovine serum albumin.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.