Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Antibody (6-5E-10B), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120909 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120909 Supplier Novus Biologicals Supplier No. NBP253103JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progesterone Monoclonal antibody specifically detects Progesterone in All samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Progesterone
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 6-5E-10B
Conjugate Janelia Fluor 549
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias 4-Pregnen-3,20-dione
Host Species Mouse
Immunogen Progesterone- 11a-hemisuccinate conjugated to bovine serum albumin (Pr11a-HMS-BSA)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.