Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Antibody (XM207), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB359752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359752 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359752 Supplier Novus Biologicals Supplier No. NB1107982AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progesterone Monoclonal antibody specifically detects Progesterone in Human samples. It is validated for ELISA

Specifications

Antigen Progesterone
Applications ELISA
Classification Monoclonal
Clone XM207
Conjugate Alexa Fluor 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 4-Pregnen-3,20-dione
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified progesterone conjugated with BSA.
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.