Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone R/NR3C3 Antibody (Alpha PR6), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1202765X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202765X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202765X Supplier Novus Biologicals Supplier No. NB1202765AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Progesterone R/NR3C3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone Alpha PR6
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P06401
Gene Alias NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Gene Symbols PGR
Host Species Mouse
Immunogen Progesterone receptor purified from chick oviduct cytosol.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5241
Test Specificity Detects the B form of the progesterone receptor (PR). This does not cross-react with estrogen receptor or glucocorticoid receptor.
Target Species Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Rabbit, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.