Learn More
Progesterone R/NR3C3 Antibody (PR484), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Progesterone R/NR3C3 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PR484 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Gene Alias | NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor |
| Gene Symbols | PGR |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human Progesterone R/NR3C3 protein (PGR) (Uniprot: P06401 ) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.