Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone R/NR3C3 Antibody (PR484), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $526.00

Specifications

Antigen Progesterone R/NR3C3
Clone PR484
Concentration 0.2mg/mL
Dilution Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page
Applications Western Blot, Immunohistochemistry (Paraffin), SDS-Page
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2333242
SDP
View Documents
Novus Biologicals
NBP2333240.02MG
0.02 mg
Each for $268.00
Only null left
Add to Cart
 
NBP23332401
SDP
View Documents
Novus Biologicals
NBP2333240.1MG
0.1 mg
Each for $526.00
Only null left
Add to Cart
 
NBP23332402
SDP
View Documents
Novus Biologicals
NBP2333240.2MG
0.2 mg N/A N/A N/A
Description

Description

Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Progesterone R/NR3C3
0.2mg/mL
Western Blot, Immunohistochemistry (Paraffin), SDS-Page
Unconjugated
Mouse
Breast Cancer, Cancer
PBS with 0.05% BSA. with 0.05% Sodium Azide
5241
A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Primary
Store at 4C.
PR484
Western Blot 0.5-1ug/ml, Simple Western 10 ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page
Monoclonal
Purified
RUO
Human
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
PGR
IgG1 κ
Protein G purified
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.