Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone R/NR3C3 Antibody (PR500) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP23332702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23332702 0.2 mg
NBP23332701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP23332702 Supplier Novus Biologicals Supplier No. NBP2333270.2MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Progesterone R/NR3C3
Applications Western Blot
Classification Monoclonal
Clone PR500
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml
Formulation 10mM PBS - BSA free with No Preservative
Gene Alias NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Gene Symbols PGR
Host Species Mouse
Immunogen progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5241
Test Specificity This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.