Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone R/NR3C3 Antibody (PR500), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120832 Supplier Novus Biologicals Supplier No. NBP233327JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Progesterone R/NR3C3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PR500
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Host Species Mouse
Immunogen Recombinant human Progesterone R/NR3C3 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5241
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.