Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone R/NR3C3 Antibody (PR501), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP233369IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233369IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233369IR Supplier Novus Biologicals Supplier No. NBP233369IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen Progesterone R/NR3C3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PR501
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Alias NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Gene Symbols PGR
Host Species Mouse
Immunogen Recombinant human Progesterone R/NR3C3 protein (Uniprot: P06401)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5241
Test Specificity This monoclonal antibody is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.