Learn More
Progesterone Receptor Ab-8 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.
Supplier: Epredia MS298PABX
Description
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.Host Species: Mouse
Clone: hPRa 2 + hPRa 3
Isotype: IgG1/κ + IgG1/κ
Species Reactivity: Human, Horse, Sheep and Pig. Others not known.
Epitope: N-terminal half of human PgR
Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (B-form) and 81kDa (A-form)
Positive Control: T47D or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Specifications
| Progesterone Receptor Ab-8 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Cancer and Tumor Biology | |
| Equine, Human, Pig, Sheep | |
| IgG2a κ |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| hPRa 2+hPRa 3 | |
| Unconjugated | |
| PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice | |
| ASR, IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.