Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progranulin/PGRN Antibody (017), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB074997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074997 Supplier Novus Biologicals Supplier No. NBP290561AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA.

Specifications

Antigen Progranulin/PGRN
Applications ELISA
Classification Monoclonal
Clone 17
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Progranulin/PGRN (NP_032201.2; Met 1-Leu 589).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2896
Test Specificity Has cross-reactivity in ELISA with: Human GRN
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.