Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP250549100UL
Description
Prohibitin 2 Polyclonal specifically detects Prohibitin 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Prohibitin 2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
PHB2 | |
KLH conjugated synthetic peptide between 106-134 amino acids from human PHB2. | |
100 μL | |
Cellular Markers, Signal Transduction | |
11331 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Western Blot, Immunohistochemistry | |
0.5 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:10 - 1:50 | |
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction