Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $610.00
Specifications
Antigen | Prohibitin 2 |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:10 - 1:50 |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Prohibitin 2 Polyclonal specifically detects Prohibitin 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Prohibitin 2 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Human, Mouse | |
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
PHB2 | |
IgG | |
Affinity Purified |
Western Blot 1:1000, Immunohistochemistry 1:10 - 1:50 | |
Polyclonal | |
Rabbit | |
Cellular Markers, Signal Transduction | |
PBS with 0.09% Sodium Azide | |
11331 | |
KLH conjugated synthetic peptide between 106-134 amino acids from human PHB2. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title