Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin 2 Rabbit anti-Human, Mouse, Rat, Clone: 8I10B1, Novus Biologicals™
SDP

Catalog No. NB168624 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168624 20 μg
NB168623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168624 Supplier Novus Biologicals Supplier No. NBP31676620UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Prohibitin 2 Monoclonal antibody specifically detects Prohibitin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Prohibitin 2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8I10B1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Prohibitin 2 (Prohibitin 2) (Q99623).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11331
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.