Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin Antibody (PHB/1882), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB391161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391161 Supplier Novus Biologicals Supplier No. NBP308723AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prohibitin Monoclonal antibody specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Prohibitin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PHB/1882
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias PHB1, prohibitin
Host Species Mouse
Immunogen Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) (Uniprot: P35232)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5245
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.