Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin Mouse anti-Human, Clone: PHB/3228, Novus Biologicals™
SDP

Catalog No. NB123188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123188 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123188 Supplier Novus Biologicals Supplier No. NBP30785720UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prohibitin Monoclonal specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Prohibitin
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PHB/3228
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias PHB1, prohibitin
Host Species Mouse
Immunogen Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5245
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.