Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ProK
GREENER_CHOICE

Catalog No. p-7250163
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Unnatural amino acid, propargyl lysine derivative

ProK is a lysine-based unnatural amino acid (UAA) used for genetic code expansion. It enables bioconjugation of fluorescent probes in E. coli or multicellular organisms.

Specifications

Chemical Name or Material N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride
CAS 1428330-91-9
Quantity 10 mg
Target Unnatural Amino Acids
Molecular Formula C10H16N2O4.HCl
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.