Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prokineticin R1/PROKR1 Antibody (420849), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312341 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312341 Supplier Novus Biologicals Supplier No. FAB4655PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prokineticin R1/PROKR1 Monoclonal specifically detects Prokineticin R1/PROKR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Prokineticin R1/PROKR1
Applications Flow Cytometry
Classification Monoclonal
Clone 420849
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias G protein-coupled receptor 73, G protein-coupled receptor ZAQ, GPR73, GPR73aG-protein coupled receptor 73, PK-R1, PKR1G-protein coupled receptor ZAQ, prokineticin receptor 1, ZAQ
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Prokineticin R1/PKR1, Met1-Lys393, Accession # Q8TCW9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10887
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.