Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin Antibody (OTI6B1), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N273609PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273609PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273609PEC7 Supplier Novus Biologicals Supplier No. NBP273609PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Prolactin
Applications Flow Cytometry
Classification Monoclonal
Clone OTI6B1
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias prolactin
Gene Symbols PRL
Host Species Mouse
Immunogen Full length recombinant protein of human Prolactin (NP_000939) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5617.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.