Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin R Antibody (T6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB30531A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30531A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30531A532 Supplier Novus Biologicals Supplier No. NB300531AF532

Mouse Monoclonal Antibody

Prolactin R Monoclonal specifically detects Prolactin R in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Prolactin R
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T6
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Gene Symbols PRLR
Host Species Mouse
Immunogen Purified rat liver PRL receptor.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5618.0
Test Specificity Detects prolactin (PRL) receptor in rat tissues. This does not cross-react with Growth Hormone (GH) receptor.
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.