Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin R Antibody (U5), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N30561PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N30561PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N30561PE594 Supplier Novus Biologicals Supplier No. NB300561PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prolactin R Monoclonal specifically detects Prolactin R in Human, Mouse, Rat, Porcine, Bovine, Equine, Hamster, Rabbit, Sheep samples. It is validated for Flow Cytometry.

Specifications

Antigen Prolactin R
Applications Flow Cytometry
Classification Monoclonal
Clone U5
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Gene Symbols PRLR
Host Species Mouse
Immunogen Purified rat liver PRL receptor.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5618.0
Target Species Human, Mouse, Rat, Pig, Bovine, Equine, Hamster, Rabbit, Sheep
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.