Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proline rich 16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17965520UL
Description
Proline rich 16 Polyclonal specifically detects Proline rich 16 in Mouse samples. It is validated for Western Blot.Specifications
Proline rich 16 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_001074693 | |
PRR16 | |
Synthetic peptide directed towards the N terminal of human Prr16. Peptide sequence GDLKDVAKELKEVVDQIDTLTSDLQLEDEMTDSSKTDTLNSSSSGTTASS. | |
Affinity Purified | |
RUO | |
51334 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DSC54, Mesenchymal stem cell protein DSC54, MGC104614, proline rich 16, proline-rich protein 16 | |
Rabbit | |
33 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction