Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proline rich 16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Proline rich 16 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17965520
![]() |
Novus Biologicals
NBP17965520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179655
![]() |
Novus Biologicals
NBP179655 |
100 μL |
Each for $487.50
|
|
|||||
Description
Proline rich 16 Polyclonal specifically detects Proline rich 16 in Mouse samples. It is validated for Western Blot.Specifications
Proline rich 16 | |
Polyclonal | |
Rabbit | |
NP_001074693 | |
51334 | |
Synthetic peptide directed towards the N terminal of human Prr16. Peptide sequence GDLKDVAKELKEVVDQIDTLTSDLQLEDEMTDSSKTDTLNSSSSGTTASS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DSC54, Mesenchymal stem cell protein DSC54, MGC104614, proline rich 16, proline-rich protein 16 | |
PRR16 | |
IgG | |
33 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title