Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proline rich 16 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31552620UL
This item is not returnable.
View return policy
Description
Proline rich 16 Polyclonal antibody specifically detects Proline rich 16 in Mouse samples. It is validated for Western BlotSpecifications
Proline rich 16 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
DSC54, Mesenchymal stem cell protein DSC54, MGC104614, proline rich 16, proline-rich protein 16 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Proline rich 16 (NP_001295016.1). | |
20 μg | |
Cell Biology | |
51334 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction