Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proline rich 16 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | Proline rich 16 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Proline rich 16 Polyclonal antibody specifically detects Proline rich 16 in Mouse samples. It is validated for Western BlotSpecifications
Proline rich 16 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
DSC54, Mesenchymal stem cell protein DSC54, MGC104614, proline rich 16, proline-rich protein 16 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Proline rich 16 (NP_001295016.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, 50% glycerol, pH7.3 | |
51334 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title