Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proline-Rich Basic Protein 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB143251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143251 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143251 Supplier Novus Biologicals Supplier No. NBP305874AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proline-Rich Basic Protein 1 Polyclonal specifically detects Proline-Rich Basic Protein 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Proline-Rich Basic Protein 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C5orf65, Chromosome 5 Open Reading Frame 65, PROB1, Weakly Similar To Basic Proline-Rich Protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Proline-Rich Basic Protein 1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389333
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.