Learn More
Promega Amino Acid Mixtures
Catalog No. p-3202720 Shop All Promega Corporation Products
Click to view available options
Product Type:
Amino Acid Mixture, Complete
Amino Acid Mixture, no Methionine
Catalog No. PR-L9961
Supplier: Promega L9961
Only null left
Add to Cart
Add to Cart
Amino Acid Mixture, Complete, is an aqueous solution containing 1mM each of the 20 essential amino acids.
  • 1mM Solution of 20 Essential Amino Acids or Combinations Thereof
  • Amino acid solution for use with Flexi(R), TnT(R) and Rabbit Reticulocyte Lysates, Wheat Germ Extract and E. coli S30 Systems
  • Choose from mix of 20 amino acids or special mixtures indicated
  • Supplied at a concentration 1mM
Specifications
Content And Storage <−65°C
Product Type Amino Acid Mixture, no Methionine
Quantity 175 μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.