Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Apo-ONE™ Homogeneous Caspase-3/7 Assay Kit

Catalog No. PRG7791 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
10 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7791 100 mL
PRG7790 10 mL
PRG7792 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG7791 Supplier Promega Supplier No. G7791
Add to Cart
Add to Cart
Request a Quote

Includes

Apo-ONE Caspase Substrate Z-DEVD-R110, 100X; Apo-ONE Homogeneous Caspase Buffer

Highly sensitive, homogeneous fluorescent apoptosis assay for use in cultured cells and with purified caspase. Just add-mix-read.

  • Fast and Sensitive Measurement of Active Caspase-3 and -7
  • Fluorescent add-mix-read apoptosis assay for use in cultured cells and with purified caspase
  • Dramatically decrease time to results with highly sensitive and simple protocol
  • Flexible assay design allows for adaptable formats and throughput needs

Specifications

Quantity 100 mL
Format Cuvettes, 96- or 384-well Plates
Content And Storage 1 x 1mL 100X Caspase substrate Z-DEVD-R110, 1 x 100mL Apo-ONE Homogeneous Caspase-3/7 Buffer. Store at -30°C to -10°C
Detection Method Fluorescence
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.