Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega AttoPhos™ AP Fluorescent Substrate System
A highly sensitive, flluorescent alkaline phosphatase substrate.
$413.85 - $1182.15
Specifications
Form | Powder |
---|---|
Substrate Type | AP (Alkaline Phosphatase) Substrate |
Storage Requirements | 4°C |
Includes: AttoPhos Substrate, AttoPhos Buffer, AttoPhos Calibration Solution (Mfr. No. S1000 only)
Description
- Sensitive Fluorescent Alkaline Phosphatase (AP) Substrate
- Low fluorescence signal until enzymatically activated
- Detection of AP to 0.1 attomole
- Linear kinetics over five orders of magnitude of AP concentration
Specifications
Powder | |
AP (Alkaline Phosphatase) Substrate | |
4°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title