Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega BL21(DE3)pLysS Competent Cells
Shop All Promega Corporation Products
Click to view available options
Quantity:
20 x 50 μL
5 x 200 μL
Description
- High-Efficiency Protein Expression from Gene with T7 Promoter and Ribosome Binding Site
- Inducible protein expression from gene with T7 RNA polymerase controlled by lac UV5 promoter
- Increased stability of expressed protein from deficiency in proteases lon and OmpT
- Lower background expression of target genes with pLysS plasmid
Specifications
Specifications
| Content And Storage | <-65°C |
| Format | Tube |
| For Use With (Application) | Allow high-efficiency Protein expression of genes under the control of a T7 promoter and having a ribosome binding site |
| Includes | Competent cell control DNA, BL21(DE3)pLysS competent cells >10 x 6cfu/μg |
| Quantity | 5 x 200 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction