Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega BTK Kinase Enzyme System

Catalog No. p-4671987 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

  • 1 x 10μg BTK Kinase Enzyme System (V2941) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant full-length human BTK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. BTK (also known as Bruton tyrosine kinase) plays a crucial role in B-lymphocyte differentiation and activation.

  • Easily Screen and Profile BTK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Both primary screening as well as Kinase selectivity profiling
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.